missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP, Chicken, Polyclonal Antibody, Abnova™
Chicken polyclonal antibody raised against recombinant Human MBP.
Brand: Abnova PAB29078.100ug
This item is not returnable.
View return policy
Specifications
| MBP | |
| Polyclonal | |
| Chicken polyclonal antibody raised against recombinant Human MBP. | |
| In PBS, pH 7.2 (0.02% sodium azide) | |
| MBP | |
| A KLH-conjugated synthetic peptide shared between the human (NP_002376, NCBI) and mouse (NP_034907, NCBI) sequences. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunocytochemistry(1:1000-1:2000) Immunohistochemistry(1:1000-1:2000) Western Blot(1:2000-1:5000) The optimal working dilution should be determined by the end user. | |
| MGC99675 | |
| Chicken | |
| Antigen affinity purification | |
| RUO | |
| 4155/17196 | |
| Store at 4°C and avoid from light. Aliquot to avoid repeated freezing and thawing. |
|
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction