missing translation for 'onlineSavingsMsg'
Learn More

MAP2, Chicken, Polyclonal Antibody, Abnova™

Product Code. 16157030
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16157030 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16157030 Supplier Abnova Supplier No. PAB29077.200ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chicken polyclonal antibody raised against recombinant Human MAP2.

Specifications

Antigen MAP2
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against recombinant Human MAP2.
Dilution Immunocytochemistry(1:1000-1:2000) Immunohistochemistry(1:1000-1:2000) Western Blot(1:2000-1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.02% sodium azide)
Gene Alias DKFZp686I2148/MAP2A/MAP2B/MAP2C
Gene Symbols MAP2
Host Species Chicken
Immunogen Two different KLH-conjugated synthetic peptides corresponding to different regions of MAP2 gene product, shared between the human (NP_002365, NCBI) and mouse (P20357, NCBI) sequences.
Purification Method Antigen affinity purification
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4133/17756
Target Species Human, Mouse
Content And Storage Store at 4°C and avoid from light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.