missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGAM, Chicken, Polyclonal Antibody, Abnova™
Chicken polyclonal antibody raised against recombinant Mouse Itgam.
Brand: Abnova PAB29076.100ug
This item is not returnable.
View return policy
Specifications
| ITGAM | |
| Polyclonal | |
| Chicken polyclonal antibody raised against recombinant Mouse Itgam. | |
| In PBS, pH 7.2 (0.02% sodium azide) | |
| CD11b/CD18/CR3/CR3A/Cd11b/F730045J24Rik/Ly-40/MAC1/Mac-1/Mac-1a | |
| Chicken | |
| Antigen affinity purification | |
| RUO | |
| 16409 | |
| Store at 4°C and avoid from light. Aliquot to avoid repeated freezing and thawing. |
|
| IgY |
| Immunocytochemistry, Immunohistochemistry | |
| Unconjugated | |
| Immunocytochemistry Immunohistochemistry The optimal working dilution should be determined by the end user. | |
| Itgam | |
| Itgam | |
| A KLH-conjugated synthetic peptide corresponding to a region near the center of the extracellular domain of Itgam gene product. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction