missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken anti-human IgG, Polyclonal, Secondary Antibody, Abnova™
Specifications
Specifications
| Antigen | Chicken anti-human IgG |
| Applications | ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Chicken polyclonal antibody raised against Human IgG. |
| Dilution | ELISA Immunocytochemistry Immunohistochemistry(1:250) Western Blot(1:1000) The optimal working dilution should be determined by the end user. |
| Formulation | In PBS, pH 7.2 (0.02% sodium azide) |
| Host Species | Chicken |
| Immunogen | Purified human IgG (whole protein). |
| Purification Method | Antigen affinity purification |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?