Learn More
HBE1, Chicken, Biotin, Polyclonal Antibody, Abnova™
Chicken polyclonal antibody against synthetic peptide of HBE1
Brand: Abnova PAB27964.50ug
Description
The epsilon globin gene (HBE) is normally expressed in the embryonic yolk sac: two epsilon chains together with two zeta chains (an alpha-like globin) constitute the embryonic hemoglobin Hb Gower I; two epsilon chains together with two alpha chains form the embryonic Hb Gower II. Both of these embryonic hemoglobins are normally supplanted by fetal, and later, adult hemoglobin. The five beta-like globin genes are found within a 45kb cluster on chromosome 11 in the following order: 5'-epsilon - G-gamma - A-gamma - delta - beta-3' [provided by RefSeq]
Sequence: AAVTSLWSKSpecifications
| HBE1 | |
| Polyclonal | |
| Biotin | |
| In Phosphate-Buffered Saline with 0.02% Sodium Azide. | |
| HBE | |
| Chicken | |
| Antigen affinity purification | |
| RUO | |
| 3046 | |
| Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles. | |
| IgY |
| Immuno-MRM (Multiple Reaction Monitoring) | |
| 1 mg/mL | |
| Chicken polyclonal antibody against synthetic peptide of HBE1 | |
| HBE1 | |
| HBE1 | |
| A Synthetic peptide corresponding to amino acids of HBE1 | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only