missing translation for 'onlineSavingsMsg'
Learn More

Chicken anti-Goat IgG, HRP, Polyclonal, Secondary Antibody, Abnova™

Product Code. 16197050
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16197050 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16197050 Supplier Abnova Supplier No. PAB29101.1mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Antigen Chicken anti-Goat IgG
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Description Chicken polyclonal antibody raised against Goat IgG. The antibody is conjugated with HRP.
Dilution ELISA Western Blot(1:5000) The optimal working dilution should be determined by the end user.
Formulation In 50mM HEPES, pH 7.0, 0.1M NaCl, (0.2% BSA, 0.01% thimerosal).
Host Species Chicken
Immunogen Purified goat IgG (whole protein) emulsified in Freund's adjuvant.
Purification Method Antigen affinity purification
Quantity 1 mL
Primary or Secondary Secondary
Target Species Goat
Content And Storage Store at 4°C and avoid from light. For long term storage dilute the antibody with sterile glycerol and store it at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.