missing translation for 'onlineSavingsMsg'
Learn More

GFP, Chicken, Polyclonal Antibody, Abnova™

Chicken polyclonal antibody against synthetic peptide of GFP

Brand:  Abnova PAB27987.100uL

Product Code. 16037877

  • £332.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

GFP
Polyclonal
Unconjugated
Western Blot (1:2000 to 1:5000) The optimal working dilution should be determined by the end user.
Chicken
Affinity purification
RUO
Human
Liquid
Immunocytochemistry, Immunohistochemistry, Western Blot
0.5 mg/mL
Chicken polyclonal antibody against synthetic peptide of GFP
In Phosphate-Buffered Saline with 0.02% Sodium Azide.
Recombinant fusion protein corresponding to full length GFP
100 μL
Primary
Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
IgY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only