Learn More
FKBP10, Chicken, Polyclonal Antibody, Abnova™
Chicken polyclonal antibody raised against native Human FKBP10.
Brand: Abnova PAB29171.10mg
Description
The protein encoded by this gene belongs to the FKBP-type peptidyl-prolyl cis/trans isomerase family. It is located in endoplasmic reticulum and acts as molecular chaperones. An alternatively spliced variant encoding different isoform has been found, but the biological validity of the variant is not determined. [provided by RefSeq
Specifications
| FKBP10 | |
| Polyclonal | |
| Chicken polyclonal antibody raised against native Human FKBP10. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| FKBP6/FKBP65/FLJ20683/FLJ22041/FLJ23833/hFKBP65 | |
| Chicken | |
| IgY Fraction | |
| RUO | |
| 60681 | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgY |
| Western Blot | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| FKBP10 | |
| FKBP10 | |
| Native purified FKBP10 from normal human third trimester placenta by reversed-order OFarrell gel electrophoresis (ROOGE). | |
| 10 mg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only