missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISP2, Chicken, Biotin, Polyclonal Antibody, Abnova™
Chicken polyclonal antibody against synthetic peptide of CRISP2
Brand: Abnova PAB27974.50ug
This item is not returnable.
View return policy
Description
Sequence: NALWHTGNTPGQVRSpecifications
| CRISP2 | |
| Polyclonal | |
| Biotin | |
| In Phosphate-Buffered Saline with 0.02% Sodium Azide. | |
| CRISP-2/GAPDL5/MGC111136/TPX1/TSP1 | |
| Chicken | |
| Antigen affinity purification | |
| RUO | |
| 7180 | |
| Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles. | |
| IgY |
| Immuno-MRM (Multiple Reaction Monitoring) | |
| 1 mg/mL | |
| Chicken polyclonal antibody against synthetic peptide of CRISP2 | |
| CRISP2 | |
| CRISP2 | |
| A Synthetic peptide corresponding to amino acids of CRISP2 | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction