Learn More
ALB Chicken anti-Human, Polyclonal Antibody, Abnova™
Chicken polyclonal antibody raised against native Human ALB.
Brand: Abnova PAB29197.500ug
Description
Albumin is a soluble, monomeric protein which comprises about one-half of the blood serum protein. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular unglycosylated serum protein of molecular weight 65,000. Albumin is synthesized in the liver as preproalbumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough endoplasmic reticulum. The product, proalbumin, is in turn cleaved in the Golgi vesicles to produce the secreted albumin. [provided by RefSeq
Specifications
| ALB | |
| Polyclonal | |
| Chicken polyclonal antibody raised against native Human ALB. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| DKFZp779N1935/PRO0883/PRO0903/PRO1341 | |
| Chicken | |
| Antigen affinity purification | |
| RUO | |
| 213 | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgY |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA Western Blot The optimal working dilution should be determined by the end user. | |
| ALB | |
| ALB | |
| Native purified normal human serum albumin. | |
| 500 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only