missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicago Sky Blue 6B, Tocris Bioscience™
Description
Potent inhibitor of L-glutamate uptake into synaptic vesicles. Allosterically inhibits macrophage migration inhibitory factor (MIF) (IC50 = 0.81 μM); inhibits MIF proinflammatory activity. Also dye used as counterstain in histochemistry.
Specifications
Specifications
| CAS | 2610-05-1 |
| Molecular Formula | C34H24N6Na4O16S4 |
| Quantity | 1 g |
| Synonym | chicago blue 6b, pontamine sky blue 6bx, brillant benzo blue 6ba, tetrasodium 4-amino-6-2-4'-2-8-amino-1-hydroxy-5,7-disulfonatonaphthalen-2-yl diazen-1-yl-3,3'-dimethoxy-1,1'-biphenyl-4-yl diazen-1-yl-5-hydroxynaphthalene-1,3-disulfonate, tetrasodium 4-amino-6-e-2-4'-e-2-8-amino-1-hydroxy-5,7-disulfonatonaphthalen-2-yl diazen-1-yl-3,3'-dimethoxy-1,1'-biphenyl-4-yl diazen-1-yl-5-hydroxynaphthalene-1,3-disulfonate |
| InChI Key | UPKAWFACSJWKND-ZXFFUEEESA-J |
| SMILES | COC1=C(C=CC(=C1)C2=CC(=C(C=C2)NN=C3C=CC4=C(C3=O)C(=C(C=C4S(=O)(=O)[O-])S(=O)(=O)[O-])N)OC)NN=C5C=CC6=C(C5=O)C(=C(C=C6S(=O)(=O)[O-])S(=O)(=O)[O-])N.[Na+].[Na+].[Na+].[Na+] |
| IUPAC Name | tetrasodium;4-amino-6-[[4-[4-[2-(8-amino-1-oxo-5,7-disulfonatonaphthalen-2-ylidene)hydrazinyl]-3-methoxyphenyl]-2-methoxyphenyl]hydrazinylidene]-5-oxonaphthalene-1,3-disulfonate |
| Molecular Weight (g/mol) | 992.79 |
| PubChem CID | 44135407 |
| Formula Weight | 992.79 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?