missing translation for 'onlineSavingsMsg'
Learn More

Chicago Sky Blue 6B (Technical Grade), TRC

Product Code. 30359253
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359253 25 g -
30368100 5 g 5g
30364615 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359253 Supplier Toronto Research Chemicals Supplier No. TRCC29197325G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2610-05-1
Molecular Formula C34H24N6Na4O16S4
Molecular Weight (g/mol) 992.79
Synonym 6,6'-[(3,3-Dimethoxy[1,1'-biphenyl]-4,4'-diyl)bis(azo)]bis[4-amino-5-hydroxy-1,3-napthalenedisulphonic Acid] Tetrasodium Salt
IUPAC Name tetrasodium;4-amino-6-[(E)-[4-[4-[(E)-(8-amino-1-hydroxy-5,7-disulfonatonaphthalen-2-yl)diazenyl]-3-methoxyphenyl]-2-methoxyphenyl]diazenyl]-5-hydroxynaphthalene-1,3-disulfonate
SMILES [Na+].[Na+].[Na+].[Na+].COc1cc(ccc1N=Nc2ccc3c(cc(c(N)c3c2O)S(=O)(=O)[O-])S(=O)(=O)[O-])c4ccc(N=Nc5ccc6c(cc(c(N)c6c5O)S(=O)(=O)[O-])S(=O)(=O)[O-])c(OC)c4

Specifications

Quantity 25 g
InChI Formula InChI=1S/C34H28N6O16S4.4Na/c1-55-23-11-15(3-7-19(23)37-39-21-9-5-17-25(57(43,44)45)13-27(59(49,50)51)31(35)29(17)33(21)41)16-4-8-20(24(12-16)56-2)38-40-22-10-6-18-26(58(46,47)48)14-28(60(52,53)54)32(36)30(18)34(22)42;;;;/h3-14,41-42H,35-36H2,1-2H3,(H,43,44,45)(H,46,47,48)(H,49,50,51)(H,52,53,54);;;;/q;4*+1/p-4
Formula Weight 991.9722
Chemical Name or Material Chicago Sky Blue 6B (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.