missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHIC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542941
This item is not returnable.
View return policy
Description
Peptide sequence: EEQLLKYSPD PVVVRGSGHV TVFGLSNKFE SEFPSSLTGK VAPEEFKASI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
This gene encodes a member of the CHIC family of proteins. The encoded protein contains a cysteine-rich hydrophobic (CHIC) motif, and is localized to vesicular structures and the plasma membrane. This gene is associated with some cases of acute myeloid leukemia.
Specifications
| CHIC2 | |
| Polyclonal | |
| Unconjugated | |
| CHIC2 | |
| 1700081B18Rik; 4930502K01Rik; BrX-like translocated in leukemia; BTL; Chic2; cysteine rich hydrophobic domain 2; cysteine-rich hydrophobic domain 2; cysteine-rich hydrophobic domain 2 protein; cysteine-rich hydrophobic domain-containing protein 2; cystein-rich hydrophobic domain 2; MGC21173; zgc:55670 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26511 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UKJ5 | |
| CHIC2 | |
| Synthetic peptide directed towards the N-terminal of human CHIC2 (aa 19-68). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction