missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHIC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542660
This item is not returnable.
View return policy
Description
Peptide sequence: LRRYAPDPVL VRGAGHITVF GLSNKFDTEF PSVLTGKVAP EEFKTSIGRV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
This gene encodes a cysteine-rich hydrophobic (CHIC) domain-containing protein, and is one of the few protein-coding genes found near the X-inactivation center. Studies in mouse indicate that the mouse ortholog of this gene is subject to X-inactivation in mouse. Experiments with other CHIC domain-containing family members show that the cysteine residues are palmitoylated post-translationally, resulting in membrane association. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| CHIC1 | |
| Polyclonal | |
| Unconjugated | |
| CHIC1 | |
| Brain X-linked protein; Brx; CHIC1; cysteine rich hydrophobic domain 1; cysteine-rich hydrophobic domain 1; cysteine-rich hydrophobic domain 1 protein; cysteine-rich hydrophobic domain-containing protein 1; cystein-rich hydrophobic domain 1; Unknown (protein for IMAGE:7449829); wu:fj33g02 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 53344 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5VXU3 | |
| CHIC1 | |
| Synthetic peptide directed towards the N-terminal of human CHIC1 (aa 81-130). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction