missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHI3L2 Polyclonal Antibody

Brand: Thermo Scientific PA5112920
This item is not returnable.
View return policy
Description
Specifications
| CHI3L2 | |
| Polyclonal | |
| Unconjugated | |
| CHI3L2 | |
| CHI3L2; CHIL2; chitinase 3 like 2; chitinase 3-like 2; chitinase-3-like protein 2; Chondrocyte protein 39; YKL39; YKL-39 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1117 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.58 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15782 | |
| CHI3L2 | |
| Recombinant Human Chitinase-3-like protein 2 protein (27-390AA). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction