missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHI3L2 Polyclonal Antibody

Sheep Polyclonal Antibody

Brand:  Invitrogen™ PA5148962

Product Code. 30534543

  • £390.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

CHI3L2 may bind glycan structure with high affinity, but not heparin. Has no chitotriosidase activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CHI3L2
Polyclonal
Unconjugated
CHI3L2
CHI3L2; CHIL2; chitinase 3 like 2; chitinase 3-like 2; chitinase-3-like protein 2; Chondrocyte protein 39; YKL39; YKL-39
Sheep
Antigen affinity chromatography
RUO
1117
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
Q15782
CHI3L2
Mouse myeloma cell line NS0-derived recombinant human Chitinase 3-like 2, Tyr27-Leu390.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.