missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHI3L1 Recombinant Rabbit Monoclonal Antibody (101), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£218.00 - £436.00

Specifications

Antigen CHI3L1
Clone 101
Concentration 1 mg/mL
Applications ELISA
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17789803
View Documents
Invitrogen™
MA543460
50 μL
£218.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17739633
GREENER_CHOICE
View Documents
Invitrogen™
MA543079
100 μL
£436.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specificity: Human CHI3L1/YKL40

Human cartilage glycoprotein 39 (GP-39), also known as YKL-40, is a glycoprotein secreted by articular chondrocytes, synoviocytes and macrophages. Serum and synovial fluid GP-39 levels are elevated in inflammatory diseases and correlate with the degree of joint destruction in rheumatoid arthritis. GP-39 is expressed in articular chondrocytes and synovial cells, as well as in liver, but is undetectable in muscle tissues, lung, pancreas, mononuclear cells and fibroblasts. GP-39 is a candidate autoantigen in rheumatoid arthritis and is important in the capacity of cells to respond to and cope with changes in their environment.
Specifications

Specifications

CHI3L1
1 mg/mL
Recombinant Monoclonal
Liquid
RUO
PBS with no preservative
38 kDa heparin-binding glycoprotein; 38kDa heparin-binding glycoprotein; 39 kDa synovial protein; 39 kDa whey protein; 40 kDa secretory signalling glycoprotein; ASRT7; AW208766; BP40; BP40 precursor; breast regression protein 39; Brp39; BRP39 protein; Cartilage glycoprotein 39; cartilage glycoprotein-39; CGP39; CGP-39; CHI3L1; Chil1; chitinase 3 like 1; Chitinase 3 like protein 1CGP 39; chitinase 3-like 1; chitinase 3-like 1 (cartilage glycoprotein-39); chitinase 3-like 1 protein; chitinase-3-like protein 1; chitinase-like 1; chitinase-like protein 1; CLP-1; DKFZp686N19119; FLJ38139; GP38K; Gp39; GP-39; hCGP39; HC-gp39; hCGP-39; HCGP-3P; Mammary gland protein 40; MGP-40; signal processing protein; signal-processing protein; SPC-40; SPS-40; YKL40; YKL-40; YYL-40
CHI3L1
IgG
Protein A
Antibody
101
ELISA
Unconjugated
Rabbit
Human
P36222
1116
Recombinant Human CHI3L1/YKL40 protein.
Primary
-20°C, Avoid Freeze/Thaw Cycles
CHI3L1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.