missing translation for 'onlineSavingsMsg'
Learn More

ChGn Antibody, Novus Biologicals™

Product Code. 18321189 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321189 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18321189 Supplier Novus Biologicals Supplier No. H00055790B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

ChGn Polyclonal antibody specifically detects ChGn in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Spécification

Antigen ChGn
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. AAH60772
Gene Alias beta4GalNAcT, Beta4GalNAcT-1, CHGN, chondroitin beta1,4 N-acetylgalactosaminyltransferase, Chondroitin beta-1,4-N-acetylgalactosaminyltransferase 1, chondroitin sulfate N-acetylgalactosaminyltransferase 1, CSGalNAcT-1, EC 2.4.1.174, FLJ11264, FLJ13760, GALNACT1
Host Species Mouse
Immunogen ChGn (AAH60772, 1 a.a. - 297 a.a.) full-length human protein. MMMVRRGLLAWISRVVVLLVLLCCAISVLYMLACTPKGDEEQLALPRANSPTGKEGYQAVLQEWEEQHRNYVSSLKRQIAQLKEELQERSEQLRNGQYQASDAAGLGLDRSPPEKTQADLLAFLHSQVDKAEVNAGVKLATEYAAVPFDSFTLQKVYQLETGLTRHPEEKPVRKDKRDELVEAIESALETLNNPAENSPNHRPYTASDFIEGIYRTERDKGTLYELTFKGDHKHEFKRLILFRPFGPIMKVENEKLNMANTLINVIVPLAKRVDKFRQFMQNFRPADEVFRCVPLSP
Purification Method IgG purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 55790
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.