missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHFR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12169-1-AP-150UL
This item is not returnable.
View return policy
Description
E3 ubiquitin-protein ligase that functions in the antephase checkpoint by actively delaying passage into mitosis in response to microtubule poisons. Acts in early prophase before chromosome condensation, when the centrosome move apart from each other along the periphery of the nucleus. Probably involved in signaling the presence of mitotic stress caused by microtubule poisons by mediating the ′Lys-48′-linked ubiquitination of target proteins, leading to their degradation by the proteasome. Promotes the ubiquitination and subsequent degradation of AURKA and PLK1. Probably acts as a tumor suppressor, possibly by mediating the polyubiquitination of HDAC1, leading to its degradation. May also promote the formation of ′Lys-63′-linked polyubiquitin chains and functions with the specific ubiquitin-conjugating UBC13-MMS2 (UBE2N-UBE2V2) heterodimer. Substrates that are polyubiquitinated at ′Lys-63′ are usually not targeted for degradation, but are rather involved in signaling cellular stress.Specifications
| CHFR | |
| Polyclonal | |
| Unconjugated | |
| CHFR | |
| CHFR, RING finger protein 196, RNF116, RNF196 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 231600, 288734, 55743 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q810L3, Q96EP1 | |
| Chfr | |
| CHFR Fusion Protein Ag2811 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction