missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals CHES, 98+%, for biochemistry

Product Code. 10404765 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10404765

Brand: Thermo Scientific Chemicals 208185000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-47-9 | C8H17NO3S | 207.288 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-47-9
Molecular Formula C8H17NO3S
Molecular Weight (g/mol) 207.288
MDL Number MFCD00003835
InChI Key MKWKNSIESPFAQN-UHFFFAOYSA-N
Synonym 2-(Cyclohexylamino)ethanesulfonic acid
PubChem CID 66898
IUPAC Name 2-(cyclohexylamino)ethanesulfonic acid
SMILES C1CCC(CC1)NCCS(=O)(=O)O

Specifications

Absorbance (0.1 M aq. soln.)
Absorbance Conditions (0.1M aq. soln.)
Melting Point 300.0°C
Color White
pH 5 to 6 (1% aq. soln.)
Physical Form Fine Crystalline Powder
Quantity 500 g
Assay Percent Range 98+%
Infrared Spectrum Authentic
Linear Formula C6H11NHCH2CH2SO3H
Packaging Plastic bottle
Merck Index 15, 2055
Solubility Information Solubility in water: 103.6g/L (20°C).
Formula Weight 207.29
Percent Purity ≥98%
Grade Biochemical
Chemical Name or Material CHES, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • CHES
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.