missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals CHES, 98+%, for biochemistry

Product Code. 10042710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10042710 100 g Plastic bottle 100g
10005923 25 g Plastic bottle 25g
10404765 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10042710 Supplier Thermo Scientific Chemicals Supplier No. 208181000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-47-9 | C8H17NO3S | 207.288 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-47-9
Molecular Formula C8H17NO3S
Molecular Weight (g/mol) 207.288
MDL Number MFCD00003835
InChI Key MKWKNSIESPFAQN-UHFFFAOYSA-N
Synonym 2-(Cyclohexylamino)ethanesulfonic acid
PubChem CID 66898
IUPAC Name 2-(cyclohexylamino)ethanesulfonic acid
SMILES C1CCC(CC1)NCCS(=O)(=O)O

Specifications

Absorbance 0.05 max. at 260nm
Absorbance Conditions (0.1M aq. soln.)
Melting Point 300.0°C
Color White
pH 5 to 6 (1% aq. soln.)
Physical Form Fine Crystalline Powder
Quantity 100 g
Assay Percent Range 98+%
Infrared Spectrum Authentic
Linear Formula C6H11NHCH2CH2SO3H
Packaging Plastic bottle
Merck Index 15, 2055
Solubility Information Solubility in water: 103.6g/L (20°C).
Formula Weight 207.29
Percent Purity ≥98%
Grade Biochemical
Chemical Name or Material CHES, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • CHES
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.