missing translation for 'onlineSavingsMsg'
Learn More

CHES, 0.5M buffer soln, pH 8.5, Thermo Scientific™

Product Code. 15484089
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15484089 100 mL 100mL
15494089 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15484089 Supplier Thermo Scientific Alfa Aesar Supplier No. J61492.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C8H17NO3S
Molecular Weight (g/mol) 207.288
MDL Number MFCD00003835
InChI Key MKWKNSIESPFAQN-UHFFFAOYSA-N
PubChem CID 66898
IUPAC Name 2-(cyclohexylamino)ethanesulfonic acid
SMILES C1CCC(CC1)NCCS(=O)(=O)O

Specifications

Concentration 0.5 M
pH 8.5
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material CHES buffer soln.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.