missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHERP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597238
This item is not returnable.
View return policy
Description
Immunogen sequence: LPAGLMAPLV KLEDHEYKPL DPKDIRLPPP MPPSERLLAA VEAFYSPPSH DRPRNSEGWE QNGLYEFFRA KMRARRRKGQ EKRNSGPSRS RSRSKSRGRS S; Positive Samples: HeLa, HepG2, A-549, Jurkat, Mouse testis; Cellular Location: Cytoplasm, Endoplasmic reticulum, perinuclear region.
CHERP is involved in calcium homeostasis, growth and proliferation.
Specifications
| CHERP | |
| Polyclonal | |
| Unconjugated | |
| CHERP | |
| 5730408I11Rik; Calcium homeostasis endoplasmic reticulum protein; CHERP; D8Wsu96e; DAN16; DAN26; ERPROT 213-21; ERPROT213-21; fc83d01; ik:tdsubc_2h12; protein with polyglutamine repeat; Scaf6; spliceosome associated factor; SCAF6; SRA1; SR-related CTD associated factor 6; SR-related CTD-associated factor 6; tdsubc_2h12; Unknown (protein for IMAGE:8005617); wu:fc83d01; xx:tdsubc_2h12; zgc:55518 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10523, 27967, 290614 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8CGZ0, Q8IWX8 | |
| CHERP | |
| A synthetic peptide corresponding to a sequence within amino acids 650-750 of human CHERP (NP_0063783). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction