missing translation for 'onlineSavingsMsg'
Learn More

CHERP, Mouse anti-Human, Clone: 2H5, Abnova™

Mouse Monoclonal Antibody

Brand:  Abnova H00010523-M01.50ug

Product Code. 16102706

  • £303.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: GSNSAPPIPDSRLGEENKGHQMLVKMGWSGSGGLGAKEQGIQDPIKGGDVRDKWDQYKGVGVALDDPYENYRRNKSYSFIARMKARDEC
Specifications

Specifications

CHERP
Monoclonal
Unconjugated
calcium homeostasis endoplasmic reticulum protein
DAN16/SCAF6/SRA1
Mouse
50 μg
Primary
Human
Antibody
ELISA, Western Blot
2H5
In 1x PBS, pH 7.4
NM_006387
CHERP
CHERP (AAH21294.1, 795 a.a. ∼ 883 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
RUO
10523
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.