Learn More
Ansell Edmont™ ChemTek™ 38-520 Series Butyl Polymer Immersion Gloves
Provides the best resistance to the most aggressive chemicals without compromising dexterity or comfort. Ansell Edmont™ ChemTek™ 38-520 Series Butyl Polymer Immersion Gloves offer superior protection against aldehydes, ketones and esters as well as concentrated mineral acids.
Brand: Ansell Edmont™ 38-520_9
This item is not returnable.
View return policy
Description
- Smooth finish in black
- Easy donning and good grip
- Maximum protection from hazardous chemicals
- Surprisingly flexible and provide excellent dexterity
- Designed for comfort with a natural, curved ergonomic shape and soft feel
- Absence of latex proteins means there's no risk of latex allergies
- Ideal for the chemical industry, oil and petrol refining, printing industry, automotive industry, mining and aerospace