missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ChemR23/CMKLR1 Monoclonal antibody specifically detects ChemR23/CMKLR1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Specifications
Specifications
| Antigen | ChemR23/CMKLR1 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | BZ194 |
| Conjugate | DyLight 488 |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CHEMERINR, chemokine receptor-like 1, chemokine-like receptor 1, CHEMR23, DEZ, G-protein coupled receptor ChemR23, G-protein coupled receptor DEZ, MGC126105, MGC126106, orphan G-protein coupled receptor, Dez |
| Host Species | Rat |
| Immunogen | This antibody was raised by immunising Wistar Furth rats with the mChemR23/CMKLR1 peptide/KLH conjugate. The immunizing amino-terminal mChemR23/CMKLR1 peptide with the sequence NH2-DSGIYDDEYSDGFGYFVDLEEASPWC-COOH, corresponding to residues residues 8-32 of mChemR23/CMKLR1, with a nonnative carboxyl-terminal cysteine to facilitate conjugation to keyhole limpet hemocyanin (KLH). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?