Learn More
ChemR23/CMKLR1 Antibody (BZ194) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NBP1-43233-0.1mg
 View more versions of this product
Product Code. 18564203
  • £388.00 / 0.10mg
Description
ChemR23/CMKLR1 Monoclonal antibody specifically detects ChemR23/CMKLR1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ChemR23/CMKLR1
Monoclonal
0.5 mg/mL
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200
P97468
Rat
Protein A or G purified
RUO
Primary
Mouse
Purified
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
BZ194
Unconjugated
PBS
CHEMERINR, chemokine receptor-like 1, chemokine-like receptor 1, CHEMR23, DEZ, G-protein coupled receptor ChemR23, G-protein coupled receptor DEZ, MGC126105, MGC126106, orphan G-protein coupled receptor, Dez
Partial synthetic peptide made to an N-terminal portion of the mouse ChemR23/CMKLR1 protein (amino acids 8-32) [UniProt P97468]
0.1 mg
GPCR, Immunology
1240
Store at 4°C. Do not freeze.
IgG2a κ