missing translation for 'onlineSavingsMsg'
Learn More

ChemR23/CMKLR1 Antibody (84939), PE/Cy7, Novus Biologicals™

Product Code. 30027555 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30027555 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30027555 Supplier Novus Biologicals Supplier No. FAB362PECY7

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ChemR23/CMKLR1 Monoclonal antibody specifically detects ChemR23/CMKLR1 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ChemR23/CMKLR1
Applications Flow Cytometry
Classification Monoclonal
Clone 84939
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias CHEMERINR, chemokine receptor-like 1, chemokine-like receptor 1, CHEMR23, DEZ, G-protein coupled receptor ChemR23, G-protein coupled receptor DEZ, MGC126105, MGC126106, orphan G-protein coupled receptor, Dez
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human ChemR23, Met1-Leu371, Accession # NP_004063
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1240
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.