missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemokine-like factor Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35678-20ul
This item is not returnable.
View return policy
Description
Chemokine-like factor Polyclonal antibody specifically detects Chemokine-like factor in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Chemokine-like factor | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| C32CKLF2, chemokine-like factor, chemokine-like factor 1, chemokine-like factor 2, chemokine-like factor 3, chemokine-like factor 4, CKLF1, CKLF3, CKLF4, HSPC224, transmembrane proteolipid, UCK-1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-67 of human Chemokine-like factor (NP_057410.1).,, Sequence:, MDNVQPKIKHRPFCFSVKGHVKMLRLVFALVTAVCCLADGALIYRKLLFNPSGPYQKKPVHEKKEVL | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51192 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction