missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX ChemiScreen™ P2Y4 Purinergic Receptor Stable Cell Line
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Description
The ChemiScreen™ P2Y4 Purinergic Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. ATP, UTP) based activation of P2RY4 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses P2RY4 alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of P2RY4 , the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Calcium Flux |
| Cell Line | 1321N1 |
| For Use With (Application) | Cell Lines |
| Primary Agonist | Uridine 5'-Triphosphate |
| Quantity | 2 Vials |
| Target Entry | P2RY4 |
| Accession Number | NM_002565 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction