missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX ChemiScreen™ MRGPRD Receptor Membrane Preparation

Product Code. 17325369
Encompass_Preferred
Click to view available options
Quantity:
1 Vial (200 units)
Unit Size:
1 ampoule
This item is not returnable. View return policy

Product Code. 17325369

Brand: DiscoverX HTS206M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The ChemiScreen™ MRGPRD Receptor Membrane Preparation measures activity of MRGPRD GPCR via GTPγS assay. Membrane preps contain 1mL of 1mg/mL frozen membranes.

The ChemiScreen™ MRGPRD Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of MRGPRD surface expression. ChemiScreen membrane preps can be used to screen for GPCR activity using GTPγS assays. Increased GPCR activation levels will lead to higher radioactive signal. Membrane preps contain 1mL of 1mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression.

Specifications

Assay Entry MRGPRD
For Use With (Application) Membrane Preps
Quantity 1 Vial (200 units)
Target Entry MRGPRD
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.