missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX ChemiScreen™ β2 Adrenergic Receptor Membrane Preparation
Click to view available options
Quantity:
1 Vial (200 units)
Unit Size:
1 ampoule
Description
The ChemiScreen™ β2 Adrenergic Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of ADRB2 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1mL of 1mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression.
Specifications
Specifications
| Assay Entry | β2 |
| For Use With (Application) | Membrane Preps |
| Quantity | 1 Vial (200 units) |
| Target Entry | ADRB2 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction