missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Chemically Defined Lipid Concentrate

Product Code. 11548846 Shop All Gibco Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11548846 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11548846 Supplier Gibco™ Supplier No. 11905031
Make Your Process Development Count
Use promo code 27978
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Chemically Defined Lipid Concentrate

Insect cell culture tested.

Chemically Defined Lipid Concentrate is a concentrated lipid emulsion designed to reduce or replace fetal bovine serum in cell culture media for a wide variety of applications, including growth and maintenance of CHO, hybridoma, and insect cells in culture; monoclonal antibody production by hybridomas; and viral expression in insect cells. The components of this medium supplement are chemically-defined.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C) and protect from light.
Cell Type Mammalian Cells
Form Liquid
Product Type Lipid Concentrate
pH 4 to 6
Quantity 100 mL
Shipping Condition Room Temperature
Do you have stability/degradation information for Chemically Defined Lipid Concentrate (Cat. No. 11905031)? How has the product been filtered?

The shelf life of the product in the closed bottle is 12 months from the date of manufacture. In order to prevent “air encounters”, we recommend keeping the head space in the bottle as small as possible. You may consider an Argon overlay as well. Argon being heavier than air will settle on top of the chemical. We recommend dividing the contents of the bottle (100 mL) in usable aliquots ideally such that each aliquot is used up in one application.

The product is filtered with a 0.22 µm Durapore filter or Cartridge from EMD Millipore (Cat. No. MSP000812).

compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diethyl chloromalonate
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: propanedioic acid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.