missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Chemical Test Kit
Click to view available options
For Use With (Application):
Acidity Determination
Ammonia Determination in Fresh Water
Ammonia Determination in Seawater
Chloride Determination
Phosphate Determination
Sulfite Determination
Packungsgröße:
Each
Beschreibung
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
- Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
- High resolution readings for fast, accurate determinations
- Replacement reagents available: No need to buy a new kit when reagents are exhausted
Spezifikation
Spezifikation
| Analytes | Mercuric Nitrate |
| For Use With (Application) | Chloride Determination |
| For Use With (Equipment) | Water samples |
| Includes | 1 (15mL) dropper bottle diphenylcarbazone indicator, 1 (30mL) dropper bottle nitric acid solution, 120mL bottle mercuric nitrate solution, 50mL calibrated vessel, 10mL calibrated vessel, calibrated syringe with tip |
| No. of Tests | 110 |
| Quantity | 1 Kit |
| Range | 0 to 100mg/L, 0 to 1000mg/L |
| Test Method | Titration |
| Type | Chloride Test Kit |
Sicherheit und Handhabung
Produktidentifikator
- 1 SET CHLORIDE TEST KIT HI4815
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
- H225-Highly flammable liquid and vapour.
- H290-May be corrosive to metals.
- H300-Fatal if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- EUH071-Corrosive to the respiratory tract.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use powder to extinguish.
- P391-Collect spillage.
- P403+P235-Store in a well-ventilated place. Keep cool.
- MIXTURE LIST-Contains : ethanol, mercury(II)nitrate, nitric acid
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur