missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Chemical Test Kit
Click to view available options
For Use With (Application):
Acidity Determination
Ammonia Determination in Fresh Water
Ammonia Determination in Seawater
Chloride Determination
Phosphate Determination
Sulfite Determination
Unit Size:
Each
Description
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
- Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
- High resolution readings for fast, accurate determinations
- Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
Specifications
| Analytes | Methyl orange / phenolphthalein |
| For Use With (Application) | Acidity Determination |
| For Use With (Equipment) | Water samples |
| Includes | 10mL dechlorinating reagent, 10mL bromophenol blue indicator, 10mL phenolphthalein indicator, 120mL acidity titrant, 10mL and 50mL calibrated vessels, calibrated syringe |
| No. of Tests | 110 |
| Quantity | 1 Kit |
| Range | 0 to 100mg/L, 0 to 500mg/L |
| Test Method | Titration |
| Type | Chemical Test Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction