Learn More
Sampling Systems™ Chemical Spoon and Spatula
Product Code. 17962012 Shop All Sampling Systems Products
Change view
Click to view available options
Length (Metric):
154 mm
201 mm
280 mm
317 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Length (Metric) unitSize
17962012 201 mm Each
17942112 154 mm Each
17952012 280 mm Each
17992012 317 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 17962012 missing translation for 'mfr' Sampling Systems™ missing translation for 'supplierNo' A625200
Il en reste null
Ajouter au panier
Ajouter au panier
This versatile chemical spoon and spatula has a robust design and is made from 304 stainless steel. It can be used as both a spoon and a spatula to allow for greater versatility in the sampling of both powder and liquid products.
  • No sharp edges or crevices, making it ideal for use in food, pharma and chemical companies
  • Available in a range of sizes
  • Heavy duty design for comfortable use
  • Ideal for measuring, sampling and dosing powders and liquids
Specifications
Sterility Non-sterile
Autoclavable Autoclavable
Disposable Reusable
Product Type Spoon
Material Stainless Steel
Length (Metric) 201 mm