Description
- No sharp edges or crevices, making it ideal for use in food, pharma and chemical companies
- Available in a range of sizes
- Heavy duty design for comfortable use
- Ideal for measuring, sampling and dosing powders and liquids
Specifications
Specifications
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Product Type | Spoon |
| Material | Stainless Steel |
| Length (Metric) | 154 mm |