missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Chemical Oxygen Demand (COD) Test Kits

Product Code. 11761043
Change view
Click to view available options
Range:
13 at 20°C
25 to 1500 mg/L
300 to 3500 mg/L
4 to 40 mg/L
6 at 20°C
9 at 20°C
Unit Size:
0.75kg
136g
Pack of 25
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range unitSize
11761043 13 at 20°C 0.75kg
11727315 25 to 1500 mg/L Pack of 25
11774725 300 to 3500 mg/L Pack of 25
11701083 4 to 40 mg/L 136g
11764725 9 at 20°C Pack of 25
10549531 6 at 20°C Pack of 25
11791073 6 at 20°C Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11761043 Supplier WTW™ Supplier No. 250359
  • ONLINE EXCLUSIVE PRICE
    £114.00
    £111.41 / 0.75kg
    Save £2.59
    2% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Get accurate Chemical Oxygen Demand (COD) test results with the Chlorine dioxide reagent test kit.

  •  Variety of instrumentation for measuring Chemical Oxygen Demand (COD)
  • Safe determinations of COD in water
  • Chlorine dioxide (ClO<sub>2</sub>) reagent test kit
  • For waste water, production control, ground water, surface water
  • For PhotoLab&reg; S6, PhotoLab&reg; S12, PhotoLab&reg; Spectral, spectroFlex photometers
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN ISO 15705
For Use With (Application) Wastewater, production control, ground water, surface water
For Use With (Equipment) PhotoLab™6, PhotoLab™ S12, PhotoLab™ 6000, PhotoLab™ Spektral photometer, Ph
No. of Tests 50
Range 13 at 20°C
Type CombiCheck Lead (Pb) reagent test kit
compliance-icons compliance-icons compliance-icons
Product Identifier
  • COD Cell Test Method: photometric Spectroquant
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: sulphuric acid, mercury(II) sulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.