Learn More
Unity Lab Services™ Chemical Filtering Hood IQOQ Service
Description
- IQ:Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order.
- Verifies equipment and any other accessories are assembled and installed. Any abnormal event(s) observed during the IQ is documented in the IQ protocol.
- Verifies that the installation site and equipment environment meet manufacturer-specified environmental requirements.
- Configuration information for each system component gets recorded.
- Installation of Equipment is NOT Included.
- OQ: System component information detailed configuration information for each system component have been recorded (as applicable).
- Verifies important equipment functions and ensures that the equipment operates as expected by the manufacturer and in conformance to standards and requirements.
- Key test varies based on relevant certification standard (NSF49/EN12469/DIN12480). Examples: filter leakage, DEHS, air velocity, electrical safety, air-flow smoke pattern, KI-DISCUS™.
Specifications
Specifications
| For Use With (Equipment) | Chemical Filtering Hood |
| Product Type | IQ/OQ Service |