missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Chemfluor FEP Tubing

Brand:  Cole-Parmer™ 96001-10

Product Code. 11735848

  • £448.00 / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Autoclavable
6.4 mm
-270°C
Sterile
213 psi
3 mm
15.24 m
1 Each
Transparent
205°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers