missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHEMetrics CHEMets Dissolved Oxygen, Visual Water Testing Kit
missing translation for 'orderingAttributeHoverText'
Range:
0 to 100 ppb
0 to 40 ppb
missing translation for 'unitSize'
Pack of 30
Beschreibung
Beinhaltet
REMOVE
Provide results in two minutes or less
- Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
- To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
- In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
- Comparators have two-year shelf life.
Spezifikation
Spezifikation
| Analytes | Dissolved Oxygen (DO) |
| Includes | Plastic case, Refill, Comparator, adhesive mounting clamp, permanent mounting clamp, sampling tube and instructions |
| No. of Tests | 30 |
| Range | 0 to 40 ppb |
| Test Time | 2 min. |
| Type | CHEMets Visual Water Testing Kit |
Sicherheit und Handhabung
Produktidentifikator
- Dissolved Oxygen Test Kit
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P363-Wash contaminated clothing before reuse.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur