Learn More
CHEMetrics CHEMets Dissolved Oxygen, Visual Water Testing Kit
Product Code. 11781004
Change view
Click to view available options
Range:
0 to 100 ppb
0 to 40 ppb
Unit Size:
Pack of 30
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
11781004 0 to 100 ppb Pack of 30
11731004 0 to 40 ppb Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11781004 Supplier CHEMetrics Supplier No. K7599
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes
REMOVE
Provides results in two minutes or less
Provide results in two minutes or less
  • Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
  • To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
  • In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
  • Comparators have two-year shelf life.
Specifications
Analytes Dissolved Oxygen (DO)
Includes Plastic case, Refill, Comparator, adhesive mounting clamp, permanent mounting clamp, sampling tube and instructions
No. of Tests 30
Range 0 to 100 ppb
Test Time 2 min.
Type CHEMets Visual Water Testing Kit
compliance-icons compliance-icons
Product Identifier
  • Dissolved Oxygen CHEMets & ULR CHEMets Refills and Vacu-vials Ampoules
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P264-Wash all exposed externalbody areas thoroughly after handling.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2, 2'-Oxydiethanol,Potassium hydroxide