missing translation for 'onlineSavingsMsg'
Learn More

CHEMetrics CHEMets Iron Test Kit Refill

Product Code. 11701044
Change view
Click to view available options
No. of Tests:
30
Unit Size:
Pack of 30
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests unitSize
11701044 30 Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11701044 Supplier CHEMetrics Supplier No. R6001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides results in two minutes or less

This product contains 30 self-filling CHEMets ampules to refill kit K-6010 for the analysis of iron (total & soluble). R-6001 is compatible with C-6001 and C-6010 comparators.

Specifications

Analytes Iron
For Use With (Equipment) C-6001 and C-6010 comparators
Includes 30 self-filling CHEMets ampoules to refill kit K-6010
No. of Tests 30
Range 0 to 1 ppm and 1 to 10 ppm
Test Method Phenanthroline
Test Time 1 min.; Total 5 min.
Type CHEMets Refill Kit
compliance-icons
Product Identifier
  • CHEMets Iron 0-10 PPM refill
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P271-Use only outdoors or in a well-ventilated area.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: acetic acid glacial,ammonium acetate, hydroxylaminehydrochloride, 1,10-phenanthroline monohydrate,
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.