missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHEMetrics CHEMets Dissolved Oxygen Test Kit Refill
Description
- Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
- To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
- In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
- Comparators have two-year shelf life
Specifications
Specifications
| Analytes | Dissolved Oxygen (DO) |
| For Use With (Equipment) | C-7540 comparators |
| Includes | 30 self-filling CHEMets ampoules to refill kits K-7540 (0-40 ppb) and K-7599 (0-100 ppb) |
| No. of Tests | 30 |
| Range | 0 to 40 ppb and 0 to 100 ppb |
| Test Method | Rhodazine D |
| Test Time | 2 min. |
| Type | CHEMets Dissolved Oxygen Refill |
Safety and Handling
Product Identifier
- Dissolved Oxygen CHEMets & ULR CHEMets Refills and Vacu-vials Ampoules
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P270-Do not eat, drink or smoke when using this product.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: 2, 2'-Oxydiethanol,Potassium hydroxide
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?