missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHEMetrics CHEMets Dissolved Oxygen Test Kit Refill
Description
- Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
- To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
- In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
- Comparators have two-year shelf life
Specifications
Specifications
| Analytes | Dissolved Oxygen (DO) |
| For Use With (Equipment) | C-7512 comparators |
| Includes | 30 self-filling CHEMets ampoules to refill kit K-7512 |
| No. of Tests | 30 |
| Range | 1 to 12 ppm |
| Test Method | Indigo Carmine |
| Test Time | 2 min. |
| Type | CHEMets Dissolved Oxygen Refill |
Safety and Handling
Product Identifier
- dissolved oxygen vacu-vials ampoules and chemets refill
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: diethylene glycol, potassium hydrogen phthalate, hydrochloric acid, EDTA disodium salt dihydrate, C.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?