missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Chemerin Monoclonal antibody specifically detects Chemerin in Mouse samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Chemerin |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 13G2 |
| Conjugate | PE |
| Dilution | ELISA |
| Formulation | PBS |
| Gene Alias | chemerin, RAR-responsive protein TIG2, retinoic acid receptor responder (tazarotene induced) 2, retinoic acid receptor responder protein 2, Tazarotene-induced gene 2 protein, TIG2HP10433 |
| Host Species | Rat |
| Immunogen | Mouse recombinant Chemerin |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?