missing translation for 'onlineSavingsMsg'
Learn More

Chemerin Antibody (13G2), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18406595 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406595 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406595 Supplier Novus Biologicals Supplier No. NBP221989MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Chemerin Monoclonal antibody specifically detects Chemerin in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chemerin
Applications ELISA
Classification Monoclonal
Clone 13G2
Conjugate mFluor Violet 500 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias chemerin, RAR-responsive protein TIG2, retinoic acid receptor responder (tazarotene induced) 2, retinoic acid receptor responder protein 2, Tazarotene-induced gene 2 protein, TIG2HP10433
Host Species Rat
Immunogen Mouse recombinant Chemerin
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5919
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.