missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemerin Antibody (13G2), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-21989CL1
This item is not returnable.
View return policy
Description
Chemerin Monoclonal antibody specifically detects Chemerin in Mouse samples. It is validated for ELISA
Specifications
| Chemerin | |
| Monoclonal | |
| CoraFluor 1 | |
| chemerin, RAR-responsive protein TIG2, retinoic acid receptor responder (tazarotene induced) 2, retinoic acid receptor responder protein 2, Tazarotene-induced gene 2 protein, TIG2HP10433 | |
| Mouse recombinant Chemerin | |
| 0.1 mL | |
| Cardiovascular Biology, metabolism | |
| 5919 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2 |
| ELISA | |
| 13G2 | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction