missing translation for 'onlineSavingsMsg'
Learn More

Chelidamic acid, MedChemExpress

Product Code. 16419386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16419386 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16419386 Supplier Medchem Express Supplier No. HYW016349

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Chelidamic acid is a heterocyclic organic acid with a pyran skeleton. Chelidamic acid has good coordination ability with noble metal ions. Chelidamic acid is also one of the most potent inhibitors of glutamate decarboxylase, with a Ki of 33 μM.

Chelidamic acid is a heterocyclic organic acid with a pyran skeleton. Chelidamic acid has good coordination ability with noble metal ions. Chelidamic acid is also one of the most potent inhibitors of glutamate decarboxylase, with a Ki of 33 μM.

Specifications

Chemical Name or Material Chelidamic acid
CAS 138-60-3
Color Brown
Purity Grade Notes Research
Molecular Formula C7H5NO5
Quantity 100 mg
Solubility Information DMSO : 10 mg/mL (54.61 mM; Need ultrasonic) ∣H2O : 1 mg/mL (5.46 mM; Need ultrasonic)
SMILES O=C(C1=CC(C=C(C(O)=O)N1)=O)O
Molecular Weight (g/mol) 183.12
Formula Weight 183.12
Percent Purity 97.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.