missing translation for 'onlineSavingsMsg'
Learn More

Chelerythrine Chloride, TRC

Product Code. 17954165
Change view
Click to view available options
Quantity:
5 mg
25 mg
250 mg
Unit Size:
250mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17954165 25 mg 25mg
17922635 5 mg 5mg
17914475 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17954165 Supplier Toronto Research Chemicals Supplier No. TRCC29188025MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3895-92-9
Molecular Formula C21 H18 N O4 . Cl
Molecular Weight (g/mol) 383.82
Synonym [1,3]Benzodioxolo[5,6-c]phenanthridinium, 1,2-dimethoxy-12-methyl-, chloride (1:1), Chelerythrine, chloride (6CI,7CI,8CI), [1,3]Benzodioxolo[5,6-c]phenanthridinium, 1,2-dimethoxy-12-methyl-, chloride (9CI), Broussonpapyrine chloride, Chelerythrinium chloride, NSC 646662
IUPAC Name 1,2-dimethoxy-12-methyl-[1,3]benzodioxolo[5,6-c]phenanthridin-12-ium;chloride
SMILES [Cl-].COc1ccc2c(c[n+](C)c3c2ccc4cc5OCOc5cc34)c1OC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H18NO4.ClH/c1-22-10-16-13(6-7-17(23-2)21(16)24-3)14-5-4-12-8-18-19(26-11-25-18)9-15(12)20(14)22;/h4-10H,11H2,1-3H3;1H/q+1;/p-1
Formula Weight 383.0924
Chemical Name or Material Chelerythrine Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.